Methylthiazol-5-carboxylat

Lieferant: COMBI-BLOCKS

HI-1115-5G HI-1115-1G
COBBHI-1115-5GEA 0 EUR
COBBHI-1115-5G COBBHI-1115-1G
Methylthiazol-5-carboxylat
Methylthiazol-5-carboxylat
Formel: C₅H₅NO₂S
MDL: MFCD03788563
CAS-Nummer: 14527-44-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR