2,5-Dibrom-1-tritylimidazol

Lieferant: COMBI-BLOCKS

HI-1126-1G
COBBHI-1126-1GEA 385 EUR
COBBHI-1126-1G
2,5-Dibrom-1-tritylimidazol
2,5-Dibrom-1-tritylimidazol
Formel: C₂₂H₁₆Br₂N₂
MDL: MFCD04115731
CAS-Nummer: 850429-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR