5-Thiazolcarbonsäure

Lieferant: COMBI-BLOCKS

HI-1129-1G HI-1129-5G
COBBHI-1129-1GEA 50.9 EUR
COBBHI-1129-1G COBBHI-1129-5G
5-Thiazolcarbonsäure
5-Thiazolecarboxylic acid
Formel: C₄H₃NO₂S
Molecular Weight: 129.1371 g/mol
MDL: MFCD03428539
CAS-Nummer: 14527-41-4
EINECS: 238-545-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR