1-Ethoxymethyl-2-iodimidazol

Lieferant: COMBI-BLOCKS

HI-1132-1G
COBBHI-1132-1GEA 193 EUR
COBBHI-1132-1G
1-Ethoxymethyl-2-iodimidazol
1-Ethoxymethyl-2-iodimidazol
Formel: C₆H₉IN₂O
Molecular Weight: 252,05 g/mol
MDL: MFCD03788568
CAS-Nummer: 146697-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR