2,5-Dibrom-1,4-dimethylimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
2,5-Dibrom-1,4-dimethyl-1H-imidazol
HI-1174-5G
HI-1174-1G
COBBHI-1174-5GEA
884
EUR
COBBHI-1174-5G
COBBHI-1174-1G
2,5-Dibrom-1,4-dimethylimidazol
2,5-Dibromo-1,4-dimethylimidazole
Formel:
C₅H₆Br₂N₂ Molecular Weight: 253.92 g/mol |
MDL:
MFCD06659900 CAS-Nummer: 850429-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...