2,5-Dibrom-1,4-dimethylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 2,5-Dibrom-1,4-dimethyl-1H-imidazol

HI-1174-5G HI-1174-1G
COBBHI-1174-5GEA 884 EUR
COBBHI-1174-5G COBBHI-1174-1G
2,5-Dibrom-1,4-dimethylimidazol
2,5-Dibromo-1,4-dimethylimidazole
Formel: C₅H₆Br₂N₂
Molecular Weight: 253.92 g/mol
MDL: MFCD06659900
CAS-Nummer: 850429-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR