Imidazol-4-carbonsäureethylester

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl imidazole-4-carboxylate

HI-1187-1G HI-1187-25G HI-1187-5G
COBBHI-1187-1GEA 0 EUR
COBBHI-1187-1G COBBHI-1187-25G COBBHI-1187-5G
Imidazol-4-carbonsäureethylester
Imidazol-4-carbonsäureethylester
Formel: C₆H₈N₂O₂
MDL: MFCD07363812
CAS-Nummer: 23785-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR