6-Bromimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HI-1235-1G HI-1235-5G
COBBHI-1235-1GEA 0 EUR
COBBHI-1235-1G COBBHI-1235-5G
6-Bromimidazo[1,2-a]pyridin
6-Bromimidazo[1,2-a]pyridin
Formel: C₇H₅BrN₂
MDL: MFCD06496200
CAS-Nummer: 6188-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR