5-Oxazolcarbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: Oxazole-5-carboxylic acid

HI-1271-1G
COBBHI-1271-1GEA 50.9 EUR
COBBHI-1271-1G
5-Oxazolcarbonsäure
5-Oxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113,07 g/mol
MDL: MFCD04114931
CAS-Nummer: 118994-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR