6-Brom-7-methylimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HI-1295-5G HI-1295-1G
COBBHI-1295-5GEA 471 EUR
COBBHI-1295-5G COBBHI-1295-1G
6-Brom-7-methylimidazo[1,2-a]pyridin
6-Brom-7-methylimidazo[1,2-a]pyridin
Formel: C₈H₇BrN₂
Molecular Weight: 211.0600 g/mol
MDL: MFCD09258769
CAS-Nummer: 116355-18-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR