8-Brom-6-methylimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HI-1296-1G HI-1296-5G
COBBHI-1296-1GEA 0 EUR
COBBHI-1296-1G COBBHI-1296-5G
8-Brom-6-methylimidazo[1,2-a]pyridin
8-Brom-6-methylimidazo[1,2-a]pyridin
Formel: C₈H₇BrN₂
MDL: MFCD09258770
CAS-Nummer: 136117-93-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR