4-Brom-1-(2-nitrophenyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1314-5G HI-1314-1G
COBBHI-1314-5GEA 208 EUR
COBBHI-1314-5G COBBHI-1314-1G
4-Brom-1-(2-nitrophenyl)-1H-pyrazol
4-Bromo-1-(2-nitrophenyl)-1H-pyrazole
Formel: C₉H₆BrN₃O₂
MDL: MFCD09475850
CAS-Nummer: 957034-96-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR