5-Methyl-2-(2-nitrophenyl)oxazol

Lieferant: COMBI-BLOCKS

HI-1330-1G
COBBHI-1330-1GEA 1690 EUR
COBBHI-1330-1G
5-Methyl-2-(2-nitrophenyl)oxazol
5-Methyl-2-(2-nitrophenyl)oxazole
Formel: C₁₀H₈N₂O₃
Molecular Weight: 204,1800 g/mol
MDL: MFCD09801026
CAS-Nummer: 52829-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR