2-Acetyl-7-brombenzofuran

Lieferant: COMBI-BLOCKS

HI-1374-5G HI-1374-1G
COBBHI-1374-5GEA 796 EUR
COBBHI-1374-5G COBBHI-1374-1G
2-Acetyl-7-brombenzofuran
2-Acetyl-7-brombenzofuran
Formel: C₁₀H₇BrO₂
Molecular Weight: 239.07 g/mol
MDL: MFCD10699679
CAS-Nummer: 460086-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR