5-Amino-4-cyan-1-(2,4-difluorphenyl)-3-methyl-1H-pyrazol
Lieferant: COMBI-BLOCKS
Synonyme:
5-Amino-1-(2,4-difluorophenyl)-3-methyl-1H-pyrazole-4-carbonitrile
HI-1401-1G
COBBHI-1401-1GEA
719
EUR
COBBHI-1401-1G
5-Amino-4-cyan-1-(2,4-difluorphenyl)-3-methyl-1H-pyrazol
5-Amino-4-cyano-1-(2,4-difluorophenyl)-3-methyl-1H-pyrazole
Formel:
C₁₁H₈F₂N₄ Molecular Weight: 234,2048 g/mol |
MDL:
MFCD08459254 CAS-Nummer: 1020057-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...