5-Amino-4-cyan-1-(2,4-dichlorphenyl)-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1402-1G
COBBHI-1402-1GEA 149 EUR
COBBHI-1402-1G
5-Amino-4-cyan-1-(2,4-dichlorphenyl)-3-methyl-1H-pyrazol
5-Amino-4-cyano-1-(2,4-dichlorophenyl)-3-methyl-1H-pyrazole
Formel: C₁₁H₈Cl₂N₄
MDL: MFCD10699683
CAS-Nummer: 58791-83-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR