5-Amino-4-cyan-1-(2,4-dimethylphenyl)-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1403-1G
COBBHI-1403-1GEA 451 EUR
COBBHI-1403-1G
5-Amino-4-cyan-1-(2,4-dimethylphenyl)-3-methyl-1H-pyrazol
5-Amino-4-cyano-1-(2,4-dimethylphenyl)-3-methyl-1H-pyrazole
Formel: C₁₃H₁₄N₄
MDL: MFCD10699684
CAS-Nummer: 1072944-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR