3-Bromimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HI-1413-5G HI-1413-1G
COBBHI-1413-5GEA 147 EUR
COBBHI-1413-5G COBBHI-1413-1G
3-Bromimidazo[1,2-a]pyridin
3-Bromimidazo[1,2-a]pyridin
Formel: C₇H₅BrN₂
Molecular Weight: 197.0300 g/mol
MDL: MFCD00014556
CAS-Nummer: 4926-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR