5-Amino-4-cyan-1-(2,5-dichlorphenyl)-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1416-1G
COBBHI-1416-1GEA 303 EUR
COBBHI-1416-1G
5-Amino-4-cyan-1-(2,5-dichlorphenyl)-3-methyl-1H-pyrazol
5-Amino-4-cyano-1-(2,5-dichlorophenyl)-3-methyl-1H-pyrazole
Formel: C₁₁H₈Cl₂N₄
MDL: MFCD10699685
CAS-Nummer: 76982-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR