5-Amino-4-cyan-1-(2-chlorphenyl)-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1417-1G
COBBHI-1417-1GEA 497 EUR
COBBHI-1417-1G
5-Amino-4-cyan-1-(2-chlorphenyl)-3-methyl-1H-pyrazol
5-Amino-4-cyan-1-(2-chlorphenyl)-3-methyl-1H-pyrazol
Formel: C₁₁H₉ClN₄
MDL: MFCD10699686
CAS-Nummer: 107856-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR