5-Amino-4-cyan-3-methyl-1-(perfluorphenyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1421-1G
COBBHI-1421-1GEA 623 EUR
COBBHI-1421-1G
5-Amino-4-cyan-3-methyl-1-(perfluorphenyl)-1H-pyrazol
5-Amino-4-cyan-3-methyl-1-(perfluorphenyl)-1H-pyrazol
Formel: C₁₁H₅F₅N₄
MDL: MFCD11053834
CAS-Nummer: 1072944-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR