5-Amino-4-cyan-1-(3-chlorphenyl)-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1422-1G
COBBHI-1422-1GEA 303 EUR
COBBHI-1422-1G
5-Amino-4-cyan-1-(3-chlorphenyl)-3-methyl-1H-pyrazol
5-Amino-4-cyan-1-(3-chlorphenyl)-3-methyl-1H-pyrazol
Formel: C₁₁H₉ClN₄
MDL: MFCD11053835
CAS-Nummer: 107842-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR