5-Amino-4-cyan-1-(2,6-dichlorphenyl)-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1424-1G
COBBHI-1424-1GEA 149 EUR
COBBHI-1424-1G
5-Amino-4-cyan-1-(2,6-dichlorphenyl)-3-methyl-1H-pyrazol
5-Amino-4-cyano-1-(2,6-dichlorophenyl)-3-methyl-1H-pyrazole
Formel: C₁₁H₈Cl₂N₄
MDL: MFCD11053837
CAS-Nummer: 1072944-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR