5-Amino-4-cyan-1-(2,5-difluorphenyl)-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1425-1G
COBBHI-1425-1GEA 497 EUR
COBBHI-1425-1G
5-Amino-4-cyan-1-(2,5-difluorphenyl)-3-methyl-1H-pyrazol
5-Amino-4-cyano-1-(2,5-difluorophenyl)-3-methyl-1H-pyrazole
Formel: C₁₁H₈F₂N₄
MDL: MFCD11053836
CAS-Nummer: 1072944-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR