5-Amino-4-cyan-3-methyl-1-(2,3,5,6-tetrafluorphenyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1447-1G
COBBHI-1447-1GEA 742 EUR
COBBHI-1447-1G
5-Amino-4-cyan-3-methyl-1-(2,3,5,6-tetrafluorphenyl)-1H-pyrazol
5-Amino-4-cyano-3-methyl-1-(2,3,5,6-tetrafluorophenyl)-1H-pyrazole
Formel: C₁₁H₆F₄N₄
MDL: MFCD11504942
CAS-Nummer: 1072944-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR