1-(4-Bromphenyl)-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 1-(4-Bromphenyl)-3-methyl-1H-pyrazol

HI-1525-1G
COBBHI-1525-1GEA 303 EUR
COBBHI-1525-1G
1-(4-Bromphenyl)-3-methyl-1H-pyrazol
1-(4-Bromophenyl)-3-methyl-1H-pyrazole
Formel: C₁₀H₉BrN₂
MDL: MFCD11848680
CAS-Nummer: 98946-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR