4-Fluor-2-mercaptobenzothiazol
Lieferant: COMBI-BLOCKS
Synonyme:
4-Fluorobenzo[d]thiazole-2(3H)-thione
HI-1549-1G
COBBHI-1549-1GEA
127
EUR
COBBHI-1549-1G
4-Fluor-2-mercaptobenzothiazol
4-Fluoro-2-mercaptobenzothiazole
Formel:
C₇H₄FNS₂ Molecular Weight: 185,25 g/mol |
MDL:
MFCD06408877 CAS-Nummer: 154327-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...