3-Iod-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Iodopyrazole

HI-1602-5G HI-1602-1G
COBBHI-1602-5GEA 0 EUR
COBBHI-1602-5G COBBHI-1602-1G
3-Iod-1H-pyrazol
3-Iodo-1H-pyrazole
Formel: C₃H₃IN₂
Molecular Weight: 193.98 g/mol
MDL: MFCD12022324
CAS-Nummer: 4522-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR