3-Iod-5-methoxy-1H-indazol

Lieferant: COMBI-BLOCKS

HI-1605-1G HI-1605-250M
COBBHI-1605-1GEA 0 EUR
COBBHI-1605-1G COBBHI-1605-250M
3-Iod-5-methoxy-1H-indazol
3-Iodo-5-methoxy-1H-indazole
Formel: C₈H₇IN₂O
MDL: MFCD07781661
CAS-Nummer: 290367-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR