3-Amino-6-brom-1H-indazol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Bromo-1H-indazol-3-amine

HI-1777-5G
COBBHI-1777-5GEA 154 EUR
COBBHI-1777-5G
3-Amino-6-brom-1H-indazol
3-Amino-6-bromo-1H-indazole
Formel: C₇H₆BrN₃
Molecular Weight: 212,05 g/mol
MDL: MFCD05665872
CAS-Nummer: 404827-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR