5-Brom-1-isopropyl-1H-indazol

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-1-isopropyl-1H-indazole

HI-1791-1G
COBBHI-1791-1GEA 310 EUR
COBBHI-1791-1G
5-Brom-1-isopropyl-1H-indazol
5-Brom-1-isopropyl-1H-indazol
Formel: C₁₀H₁₁BrN₂
MDL: MFCD18783165
CAS-Nummer: 1280786-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR