Indol-4-carbonitril

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 4-Indolcarbonitril, 4-Cyanindol, 1H-Indol-4-carbonitril, 4-Cyan-1H-indol

IN-0005-5G IN-0005-25G IN-0005-1G
COBBIN-0005-5GEA 37.9 EUR
COBBIN-0005-5G COBBIN-0005-25G COBBIN-0005-1G
Indol-4-carbonitril
Indol-4-carbonitril
Formel: C₉H₆N₂
Molecular Weight: 142.16 g/mol
MDL: MFCD00152045
CAS-Nummer: 16136-52-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR