6-Indolcarbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
1H-Indole-6-carboxylic acid
IN-0018-100G
IN-0018-25G
IN-0018-5G
COBBIN-0018-100GEA
0
EUR
COBBIN-0018-100G
COBBIN-0018-25G
COBBIN-0018-5G
6-Indolcarbonsäure
6-Indolcarbonsäure
Formel:
C₉H₇NO₂ Molecular Weight: 161.16 g/mol Schmelzpunkt: 254…257 °C |
MDL:
MFCD00210441 CAS-Nummer: 1670-82-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...