6-Bromindol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Bromo-1H-indole

IN-0045-1G IN-0045-25G IN-0045-100G IN-0045-5G
COBBIN-0045-1GEA 0 EUR
COBBIN-0045-1G COBBIN-0045-25G COBBIN-0045-100G COBBIN-0045-5G
6-Bromindol
6-Bromindol
Formel: C₈H₆BrN
Lagertemperatur: Raumtemperatur
MDL: MFCD00238550
CAS-Nummer: 52415-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR