6-Methoxyindol

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 6-Methoxy-1H-indol, Indol-6-ylmethylether

IN-0054-5G IN-0054-25G IN-0054-1G
COBBIN-0054-5GEA 142 EUR
COBBIN-0054-5G COBBIN-0054-25G COBBIN-0054-1G
6-Methoxyindol
6-Methoxyindol
Formel: C₉H₉NO
Molecular Weight: 147.18 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00022780
CAS-Nummer: 3189-13-7
EINECS: 221-689-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR