4-Benzyloxyindol
Lieferant: COMBI-BLOCKS
IN-0056-5G
IN-0056-25G
COBBIN-0056-5GEA
0
EUR
COBBIN-0056-5G
COBBIN-0056-25G
4-Benzyloxyindol
4-Benzyloxyindol
Formel:
C₁₅H₁₃NO |
MDL:
MFCD00047200 CAS-Nummer: 20289-26-3 EINECS: 243-690-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...