4-Benzyloxyindol

Lieferant: COMBI-BLOCKS

IN-0056-5G IN-0056-25G
COBBIN-0056-5GEA 0 EUR
COBBIN-0056-5G COBBIN-0056-25G
4-Benzyloxyindol
4-Benzyloxyindol
Formel: C₁₅H₁₃NO
MDL: MFCD00047200
CAS-Nummer: 20289-26-3
EINECS: 243-690-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR