Methyloxindol-7-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-oxoindoline-7-carboxylate

IN-0089-1G
COBBIN-0089-1GEA 247 EUR
COBBIN-0089-1G
Methyloxindol-7-carboxylat
Methyloxindol-7-carboxylat
Formel: C₁₀H₉NO₃
Molecular Weight: 191,19 g/mol
MDL: MFCD02179610
CAS-Nummer: 380427-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR