4,5-Difluoroxindol

Lieferant: COMBI-BLOCKS

Synonyme: 4,5-Difluoroindolin-2-one

IN-0101-1G
COBBIN-0101-1GEA 459 EUR
COBBIN-0101-1G
4,5-Difluoroxindol
4,5-Difluoroxindol
Formel: C₈H₅F₂NO
Molecular Weight: 169,13 g/mol
MDL: MFCD04112481
CAS-Nummer: 850429-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR