Methyloxindol-6-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 2-Oxoindoline-6-carboxylate, 2-Oxoindoline-6-carboxylic acid methyll ester
IN-0107-1G
COBBIN-0107-1GEA
25.9
EUR
COBBIN-0107-1G
Methyloxindol-6-carboxylat
Methyloxindol-6-carboxylat
Formel:
C₁₀H₉NO₃ Molecular Weight: 191,1800 g/mol |
MDL:
MFCD03095196 CAS-Nummer: 14192-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...