Oxindol-6-carbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Oxoindoline-6-carboxylic acid
IN-0116-1G
COBBIN-0116-1GEA
262
EUR
COBBIN-0116-1G
Oxindol-6-carbonsäure
Oxindol-6-carbonsäure
Formel:
C₉H₇NO₃ Molecular Weight: 177,1600 g/mol |
MDL:
MFCD01415801 CAS-Nummer: 334952-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...