6-Brom-2-mercapto-4-(trifluormethyl)benzothiazol

Lieferant: COMBI-BLOCKS

IN-0216-5G
COBBIN-0216-5GEA 149 EUR
COBBIN-0216-5G
6-Brom-2-mercapto-4-(trifluormethyl)benzothiazol
6-Bromo-2-mercapto-4-(trifluoromethyl)benzothiazole
Formel: C₈H₃BrF₃NS₂
MDL: MFCD15143580
CAS-Nummer: 1215206-27-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR