5-Brom-3-iod-1H-indazol

Lieferant: COMBI-BLOCKS

IN-0271-1G
COBBIN-0271-1GEA 56.9 EUR
COBBIN-0271-1G
5-Brom-3-iod-1H-indazol
5-Bromo-3-iodo-1H-indazole
Formel: C₇H₄BrIN₂
Molecular Weight: 322,9300 g/mol
MDL: MFCD07781637
CAS-Nummer: 459133-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR