2,4-Dichlor-5-fluorpyrimidin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluoro-2,4-dichloropyrimidine

OR-0140-100G
COBBOR-0140-100GEA 232 EUR
COBBOR-0140-100G
2,4-Dichlor-5-fluorpyrimidin
2,4-Dichlor-5-fluorpyrimidin
Formel: C₄HCl₂FN₂
Molecular Weight: 166,97 g/mol
MDL: MFCD00233551
CAS-Nummer: 2927-71-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR