DL-Prolin

Lieferant: COMBI-BLOCKS

Synonyme: (±)-Prolin, (±)-1,3,4,5-Tetrahydro-2H-pyrrol- 2-carbonsäure, (±)-Pyrrolidin-2-carbonsäure, (±)-Azolidin-2-carbonsäure, H-DL-Pro-OH

OR-0258-100G
COBBOR-0258-100GEA 127 EUR
COBBOR-0258-100G
DL-Prolin
DL-Prolin
Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
Siedepunkt: 245 °C (1013 hPa)
Schmelzpunkt: 208…210 °C
Dichte: 1,38 g/cm³ (22.9 °C)
MDL: MFCD00005250
CAS-Nummer: 609-36-9
EINECS: 210-189-3
Merck Index: 12,07963

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR