(S)-(-)-4-Benzyl-2-oxazolidinon

Lieferant: COMBI-BLOCKS

Synonyme: (S)-4-Benzyl-2-oxazolidinone, (S)-4-Benzyloxazolidin-2-one

OR-0265-500G
COBBOR-0265-500GEA 370 EUR
COBBOR-0265-500G
(S)-(-)-4-Benzyl-2-oxazolidinon
(S)-(-)-4-Benzyl-2-oxazolidinon
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
MDL: MFCD00064496
CAS-Nummer: 90719-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR