Benzophenonimin
Lieferant: COMBI-BLOCKS
Synonyme:
Benzophenoneimine
OR-0525-100G
OR-0525-25G
COBBOR-0525-100GEA
137
EUR
COBBOR-0525-100G
COBBOR-0525-25G
Benzophenonimin
Benzophenonimin
Formel:
C₁₃H₁₁N Molecular Weight: 181.24 g/mol |
MDL:
MFCD00001760 CAS-Nummer: 1013-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...