Benzophenonimin

Lieferant: COMBI-BLOCKS

Synonyme: Benzophenoneimine

OR-0525-100G OR-0525-25G
COBBOR-0525-100GEA 137 EUR
COBBOR-0525-100G COBBOR-0525-25G
Benzophenonimin
Benzophenonimin
Formel: C₁₃H₁₁N
Molecular Weight: 181.24 g/mol
MDL: MFCD00001760
CAS-Nummer: 1013-88-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR