3-Nitrophthalonitril

Lieferant: COMBI-BLOCKS

OR-0559-5G
COBBOR-0559-5GEA 0 EUR
COBBOR-0559-5G
3-Nitrophthalonitril
3-Nitrophthalonitril
Formel: C₈H₃N₃O₂
Schmelzpunkt: 160 °C
MDL: MFCD00191558
CAS-Nummer: 51762-67-5
UN: 3276
ADR: 6.1,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR