HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)

Lieferant: COMBI-BLOCKS

OR-0618-100G OR-0618-25G
COBBOR-0618-100GEA 0 EUR
COBBOR-0618-100G COBBOR-0618-25G
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
Formel: C₁₀H₁₅F₆N₆OP
MDL: MFCD00274639
CAS-Nummer: 148893-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR